Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, HPLC Grade, Ricca Chemical
SDP

Catalog No. RSOA00204C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOA00204C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOA00204C Supplier Ricca Chemical Company Supplier No. RSOA00204C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100
Appearance Colorless Liquid
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.0003%
Titratable Acid max. 8 μeq/g
Titratable Base max. 0.6 μeq/g
Water max. 0.02%
Boiling Point 82°C
Grade HPLC
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.