Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, LC-MS Grade, 99.8%

Catalog No. AA47138M1
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47138M1 4 x 1 L
AA47138K2 1 L
AA47138K7 4 L
AA47138M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA47138M1 Supplier Thermo Scientific Chemicals Supplier No. 047138M1
Only null left
Add to Cart

Solvent for LC-MS analysis | CAS: 75-05-8 | C2H3N | 41.053 g/mol

Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C to -45°C
Color Colorless
Density 0.786 g/mL
Quantity 4 x 1 L
Flash Point 5°C (41°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
UN Number UN1648
Beilstein 741857
Merck Index 14,70
Boiling Point 80°C to 82°C
Solubility Information Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons.
Formula Weight 41.05
Percent Purity 99.8%
Grade LC-MS
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber 200-835-2
RTECSNumber AL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.