Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, HPLC/Spectro Grade, ≥99.9%, MP Biomedicals™

Catalog No. MP230000816 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP230000816 4 x 4 L
MP230000804 4 L
MP230000801 1 L
MP230000806 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP230000816 Supplier MP Biomedicals, Inc Supplier No. 0230000816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetonitrile is widely used as a polar aprotic solvent in organic synthesis and in the purification of butadiene in refineries.

  • Acetonitrile also plays an important role as the main solvent used in the manufacture of DNA oligonucleotides from monomers.
  • This solvent is an ideal choice as a mobile phase in HPLC for a variety of reasons.
  • Not only is it miscible with water and with many other organic solvents, but it also has a lower viscosity and produces less pressure in the column than other solvents such as Methanol.
  • The inherent low absorbance of Acetonitrile (especially HPLC grade) makes it an ideal solvent for the mobile phase of HPLC due to the reduction in the amount of noise during UV detection.
  • Overall, Acetonitrile has many unique properties as an organic mobile phase that often result in better chromatographic peaks.
  • This colorless liquid is the simplest organic nitrile (hydrogen cyanide is a simpler nitrile, but the cyanide anion is not classed as organic).
  • It is produced mainly as a byproduct of acrylonitrile manufacture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym Methyl Cyanide, Cyanomethane, Ethanenitrile
PubChem CID 6342
ChEBI CHEBI:38472

Specifications

Melting Point -49°F
Color Clear
Density 0.786 g/cm3 (at 20°C)
Quantity 4 x 4 L
Flash Point 2°C (35.6°F)
Physical Form Liquid
Refractive Index 20/D 1.344
Titratable Acid <0.008 meq/g
Titratable Base <0.0006 meq/g
Vapor Pressure 94.51 to 98.64 hPa (at 20°C)
Water ≤0.02%
UN Number UN1648
Beilstein 741857
Boiling Point 81°C to 82°C
Solubility Information Soluble in water.
SMILES CC#N
IUPAC Name acetonitrile
Grade ≥99.9%
Show More Show Less
storageNote1 Room Temperature, store under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.