Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, OmniSolv™, Non−UV, MilliporeSigma™

Catalog No. max01551 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle (Pre-packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01551 4 L Glass Bottle (Pre-packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX01551 Supplier MilliporeSigma Supplier No. AX01551
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Quantity 4 L
Flash Point 2°C
Packaging Glass Bottle (Pre-packed)
Physical Form Clear liquid
Titratable Acid 8.3μeq/g (max)
Vapor Pressure 97 hPa (20°C)
Water 0.10% max
Boiling Point 81.6°C (1013 hPa)
Formula Weight 41.05
Percent Purity 99.9% min
Chemical Name or Material Acetonitrile, Non UV
Show More Show Less
EINECSNumber 200-835-2
RTECSNumber AL7700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.