Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Supelco

Catalog No. 11100264 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-264 2 L
11-100-252 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-264 Supplier MilliporeSigma Supelco Supplier No. 1037252002
Add to Cart
Add to Cart

For UHPLC-MS LiChrosolv™

  • Intended for U/HPLC and LC-MS/MS applications.
  • Featured as a solvent in screening of benzofurans and ethylphenidate psychostimulants in serum, urine, and hair samples by UHPLC-MS/MS.
  • The product is an organic solvent, used as a mobile phase/extraction solvent in the ultra-high performance liquid chromatorgraphy - tandem mass spectrometry (UHPLC-MS/MS) analysis of pesticides in tobacco samples.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.05

Specifications

Quantity 2 L
Boiling Point 81 to 82°C (lit.)
Percent Purity Unknown
Grade Ungraded
Chemical Name or Material Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.