Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonylacetone 97.0+%, TCI America™
SDP

Catalog No. H0101100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0101100ML 100 mL
H010125ML 25 mL
H0101500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0101100ML Supplier TCI America Supplier No. H0101100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Specifications

Melting Point -9°C
Boiling Point 188°C
Quantity 100 mL
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetonylacetone
EINECSNumber (2)-0563
RTECSNumber MO3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.