Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone, 98%, pure

Catalog No. AC102410025
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102410025 2.5 L Glass bottle
AC102410050 5 mL Glass bottle
AC102412500 250 mL Glass bottle
AC102410010 1 L Glass bottle
AC102410100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC102410025 Supplier Thermo Scientific Chemicals Supplier No. 102410025
Add to Cart

CAS: 98-86-2 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-86-2
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 19.6°C
Color Colorless to Yellow
Density 1.0280g/mL
Boiling Point 202.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8O
Refractive Index 1.5315 to 1.5340
Linear Formula C6H5COCH3
Quantity 2.5 L
Beilstein 07, IV, 619
Fieser 04,5
Specific Gravity 1.028
Merck Index 15, 71
Solubility Information Solubility in water: 5.5g/L (20°C). Other solubilities: soluble in conc. h2so4,freely soluble in alcohol,chloroform,ether and,glycerol
Molecular Weight (g/mol) 120.15
Viscosity 1.8 mPa.s (20°C)
Formula Weight 120.15
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Acetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 202-708-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.