Learn More
Acetophenone Solution, 2,000 μg/mL in Methylene Chloride, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| CAS | 98-86-2 |
| Linear Formula | C8H8 O |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 120.15 |
| Grade | Certified Reference Material |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in methylene chloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.