Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Acetosyringone, Phenolic Compound, Powder, >98% Purity, 1 g

Catalog No. 502704588
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4588 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4588 Supplier Abcam Supplier No. AB1465331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naturally occurring phenolic compound that acts as an effective inducer of Agrobacterium tumefaciens virulence genes.

  • Molecular Weight: 196.2 g/mol
  • Purity: >98%
  • Form: Powder
  • CAS Number: 2478-38-8
  • Key Features: Naturally occurring phenolic compound; inducer of Agrobacterium tumefaciens virulence genes; utilized in the synthesis of anticancer agents in vitro
  • Storage: Store at ambient temperature

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Format Vial
Form Powder
Substrate Properties Chemical Substrate
Target Molecule Virulence Genes (Agrobacterium tumefaciens)
Buffer 100% Acetosyringone
Shipping Condition Ambient
Formulation 100% Acetosyringone
Quantity 1 g
For Use With (Application) Plant-pathogen recognition, Agrobacterium tumefaciens virulence gene induction, anticancer agent research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.