Learn More
CPC Scientific Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RENI005A
Encompass
Renin substrate.ONE-LETTER SEQUENCE: Ac-DRVYIHPFHLLVYSMOLECULAR FORMULA: C87H125N21O21MOLECULAR WEIGHT:1801.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [66641-26-7], RESEARCH AREA: Cardiovascular
Catalog No. 50-211-0361