Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Acetyl-calpastatin 184-210 (human)

Catalog No. 29501
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-501 Supplier Tocris Bioscience™ Supplier No. 2950/1
This item is not returnable. View return policy

Selective calpain inhibitor

Acetyl-Calpastatin (184-210) (human) is a selective calpain inhibitor. Strongly inhibits calpain I (Ki = 0.2 nM) and II but does not inhibit papain, trypsin and cathepsin L (Ki = 6 μM). Increases secretion of amyoid β-protein (Aβ) 42, Aβ40 and Aβ42 ratio.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.