Learn More
MilliporeSigma™ Acetyl-Histone H3 Immunoprecipitation (ChIP) Assay Kit
Catalog No. 17245 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Assays/Kit
Catalog No. Quantity
17-245 22 Assays/Kit
1 options
Catalog No. 17-245 Supplier MilliporeSigma™ Supplier No. 17245
Add to Cart
Add to Cart
Includes
  • Anti-acetyl-Histone H3
  • Protein A agarose/Salmon Sperm DNA
  • All necessary buffers
Used to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-Acetyl-Histone H3
  • For use to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-Acetyl-Histone H4, ChIP grade rabbit antiserum
  • Detection of the gene or promoter of interest in immunoprecipitated chromatin using quantitative PCR or Southern slot-blot analysis
TRUSTED_SUSTAINABILITY
Specifications
Quantity 22 Assays/Kit