Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Acetyl-L-alanyl-L-glutamine, Polypeptide, Solid, 500 mg

Catalog No. 502705147
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5147 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5147 Supplier Abcam Supplier No. AB145520500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypeptide for proteomics research with >98% purity.

  • Molecular Weight: 259.26 Da
  • Purity: >98%
  • Form: Solid
  • Chemical Name: (2S)-2-[[(2S)-2-acetamidopropanoyl]amino]-5-amino-5-oxopentanoic acid
  • CAS Number: 121574-43-4
  • Storage: Store at ambient temperature

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Substrate Properties Peptide-based Substrate
Target Molecule Acetyl-L-alanyl-L-glutamine
Formulation 100% (2S)-2-[[(2S)-2-acetamidopropanoyl]amino]-5-amino-5-oxopentanoic acid
Quantity 500 mg
For Use With (Application) Proteomics research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.