Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Acetyl-L-alanyl-L-glutamine, Polypeptide, Solid, Purity >98%, 5 g

Catalog No. 502705146
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5146 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5146 Supplier Abcam Supplier No. AB1455205G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity polypeptide for proteomics research.

  • Molecular Weight: 259.26 Da
  • Purity: >98%
  • Form: Solid
  • CAS Number: 121574-43-4
  • Composition: 100% (2S)-2-[[(2S)-2-acetamidopropanoyl]amino]-5-amino-5-oxopentanoic acid
  • Application: Polypeptide for proteomics research

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Substrate Properties Peptide-based Substrate
Target Molecule Acetyl-L-alanyl-L-glutamine
Formulation 100% (2S)-2-[[(2S)-2-acetamidopropanoyl]amino]-5-amino-5-oxopentanoic acid
Quantity 5 g
For Use With (Application) Proteomics research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.