Learn More
Description
Acetyl Salicylic Acid USP comes as either white crystals, commonly tabular or needle-like, or as white crystalline powder. It can be odorless to havings a faint odor. Acetyl Salicylic Acid is stable in dry air; in moist air it gradually hydrolyzes to salicylic and acetic acids. It is slightly hygroscopic.
Alternative name(s): Aspirin, Acetylsalicylic acid, Salicylic acid acetate
Specifications
Specifications
| Chemical Name or Material | Acetyl Salicylic Acid USP |
| Packaging | Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 50-78-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 180.1582 |
| Quantity | 500 g |
| Percent Purity | 99.5 to 100.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
