Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acetyl-SMC2 (Lys114) Monoclonal Antibody (KT183)
GREENER_CHOICE

Catalog No. PIMA549500
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA549500 100 μL
1 options

Catalog No. PIMA549500

Supplier: Invitrogen™ MA549500

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product was used in an Indirect ELISA.

CAP-E and CAP-C (Chromosome Associated Protein-E & C) are also SMC (Structural Maintenance of Chromosome) family members that form a heterodimeric complex required for mitotic chromosome condensation to achieve proper segregation of genetic information during subsequent cell division. hCAP-C/hCAP-E is a component of a multiprotein complex called condensin that interacts with phosphorylation of Histone H3 resulting in the mitotic chromosome condensation. The distribution patterns of the two heterodimeric complexes in interphase nucleus indicate independent behavior of the two complexes during cell cycle. These results suggest that the two distinct complexes are involved in different aspects of mitotic chromosome organization in humar cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-SMC2 (Lys114)
Applications Immunoprecipitation, Western Blot, Western Blot
Classification Monoclonal
Clone KT183
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SMC2
Gene Accession No. O95347
Gene Alias 5730502P04Rik; AI255214; AW545314; CAP E; CAPE; CAP-E; Chromosome-associated protein E; FGF-inducible protein 16; fibroblast growth factor inducible 16; Fin16; FLJ10093; hCAP-E; PRO0324; SMC protein 2; Smc2; SMC-2; SMC2 (structural maintenance of chromosomes 2, yeast)-like 1; SMC2 protein; SMC2 structural maintenance of chromosomes 2-like 1; Smc2l1; structural maintenance of chromosomes (SMC) family member, chromosome-associated protein E; structural maintenance of chromosomes 2; structural maintenance of chromosomes protein 2; XCAP-E homolog
Gene Symbols SMC2
Host Species Mouse
Immunogen Human SMC2 peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.