Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylacetone, Reagent Grade, ≥99%, Solstice

Catalog No. 60048142 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-142 2.5 L Glass bottle
60-048-143 250 mL Glass bottle
60-048-141 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 60-048-142 Supplier Solstice Supplier No. P77542.5L
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Color Colorless
Density 0.975g/mL at 25°C
pH 6 (at 200.00g/L)
Boiling Point 139.5°C (1013 hPa)
Flash Point 38°C (100.4°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3COCH2COCH3
Quantity 2.5 L
UN Number UN2310
Beilstein 741937
Solubility Information 153.8 - 155.2g/L at 20°C in water
Vapor Density 3.5 (vs air)
Formula Weight 100.12g/mol
Percent Purity ≥99%
Chemical Name or Material Acetylacetone
Show More Show Less
Hazard Statement P210-P261-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378
DOTInformation 6.1(3)/PGIII
RTECSNumber 204-634-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.