Learn More
Abcam Acetylated Alpha Tubulin Peptide, Synthetic, Human, 1 mg
Description
- Protein Type: Synthetic
- Purity: ≥70%
- Formulation: Liquid, pH: 6.75
- Constituents: 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
- Tested Applications: BL
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Acetylated Alpha Tubulin (Human) |
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Content And Storage | Store at -20°C. |
| Concentration | 1 mg/mL |
| Form | Liquid |
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75 |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.