Learn More
Bioworld Bovine Serum Albumin (BSA), Acetylated, 500 mg
Supplier:  Bioworld 220700001
Encompass_Preferred
Acetylated to inactivate nucleases commonly found in BSA. Acetylated BSA should not be used as a protein standard since acetylation of tyrosine residues prevents color development in the Lowry assay and other similar protein determinations.It is prepared by a modified method of the Cohn cold ethanol fractionation method.grade: for molecular biologysuitability: suitable for blocking Southern blotsforeign activity: DNase, RNase, protease, none detectedAppearance: lyophilized powderHS Code : 350290700PH Value: 6.5-7.5 (1% in 0.15 M sodium chloride)
Catalog No. 50-253-884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.