Learn More
Bioassay Systems EnzyChrom™ Acetylcholine Assay Kit. For quantitative determination of acetylcholine and evaluation of drug effects on acetylcholine metabolism.
SDP
Catalog No. 50489241 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of acetylcholine and evaluation of drug effects on acetylcholine metabolism. Key Features: Use 20 µL samples. Linear detection range: colorimetric assay 10 to 200 µM, fluorimetric assay 0.4 to 10 µM acetylcholine. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, milk, tissue, and cell culture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 10, 0.4 µM.
Catalog No. 50-489-241 Supplier Bioassay Systems Supplier No. EACL100
For quantitative determination of acetylcholine and evaluation of drug effects on acetylcholine metabolism. Key Features: Use 20 µL samples. Linear detection range: colorimetric assay 10 to 200 µM, fluorimetric assay 0.4 to 10 µM acetylcholine. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, milk, tissue, and cell culture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: OD, FL: 10, 0.4 µM.