Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetylcholine Chloride Solution, 0.01%
SCI_ED

Catalog No. s25652 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
0.01%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25652 100 mL 0.01%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25652

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 60-31-1 , 7732-18-5
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Density ∽1g/mL
Packaging 0.01%
Quantity 100 mL
Chemical Name or Material Acetylcholine Chloride Solution, 0.01%
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.