Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Acetylcholinesterase Assay Kit, Fluorometric, 0.1 mU/mL Sensitivity, 200 Tests

Catalog No. 502583475
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3475 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3475 Supplier Abcam Supplier No. AB138872200TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay kit designed for the rapid quantification of acetylcholinesterase activity.

  • Assay type: Enzyme activity
  • Detection method: Fluorescent
  • Sensitivity: 0.1 mU/mL
  • Sample types: Cell culture extracts
  • Applications: Enzyme activity assay
  • Features: Includes AChE positive control

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Acetylcholinesterase
For Use With (Equipment) Fluorometric Plate Reader
Detection Method Fluorescence
Detection Location In-Solution Detection
Format Kit
No. of Reactions 200 Tests
Sample Type Cell culture extracts
Quantity 200 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.