Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Acetylcysteine, USP (N-Acetyl-L-Cysteine) (Dietary Supplement Grade)
SDP

Catalog No. 14185637 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-637 25 g
14-185-634 500 g
14-185-271 1 kg
14-185-560 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-185-637 Supplier Medisca Supplier No. 049504
Add to Cart
Add to Cart

Acetylcysteine, USP (N-Acetyl-L-Cysteine) (Dietary Supplement Grade) comes as either a white to almost white crystalline powder or as colourless crystals, having a slight odor.

Acetylcysteine USP comes as either a white to almost white crystalline powder or as colourless crystals, having a slight odor.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): NAC, N-Acetylcysteine

Specifications

Chemical Name or Material Acetylcysteine, USP (N-Acetyl-L-Cysteine) (Dietary Supplement Grade)
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 616-91-1
Type Drug Standard
Molecular Weight (g/mol) 163.2
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.