Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals

Catalog No. AAJ64892LB0
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64892LB0 5 mg
AAJ64892MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64892LB0 Supplier Thermo Scientific Chemicals Supplier No. J64892LB0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

A carboxyrhodamine 110 based labeling reagent

A carboxyrhodamine 110 based labeling reagent; reacts with azides via a copper-catalyzed click reactions. Carboxyrhodamine 110 is a bright and photostable green fluorescent label that can be excited at a wavelength of 488nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1615739-95-1
Molecular Formula C32H33N3O8
MDL Number MFCD22380741
Synonym Acetylene-Fluor 488; Rhodamine 110-PEG4-alkyne

Specifications

Color Red
Quantity 5 mg
Solubility Information Soluble in DMSO,DMF,or methanol
Formula Weight 587.63
Physical Form Solid
Chemical Name or Material Acetylene-PEG4-carboxyrhodamine 110 conjugate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.