Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylenedicarboxylic Acid Monopotassium Salt 95.0+%, TCI America™
SDP

Catalog No. A083025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A083025G 25 g
A0830500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A083025G Supplier TCI America Supplier No. A083025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 928-04-1
Molecular Formula C4HKO4
Molecular Weight (g/mol) 152.146
MDL Number MFCD00013143
InChI Key KLLYWRUTRAFSJT-UHFFFAOYSA-M
Synonym acetylenedicarboxylic acid monopotassium salt, 2-butynedioic acid, monopotassium salt, potassium hydrogen butynedioate, potassium 3-carboxyprop-2-ynoate, 2-butynedioic acid, potassium salt 1:1, acetylenedicarboxylic acid, monopotassium salt, monopotassium butynedioate, potassium hydrogen acetylenedicarboxylate, monopotassium salt of acetylenedicarboxylic acid, monopotassium acetylenedicarboxylate
PubChem CID 23677964
IUPAC Name potassium;4-hydroxy-4-oxobut-2-ynoate
SMILES C(#CC(=O)[O-])C(=O)O.[K+]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 152.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetylenedicarboxylic Acid Monopotassium Salt
EINECSNumber (2)-3394
RTECSNumber AP0700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.