Learn More
Cayman Chemical AcetylharpagIde 10mg
SDP
Supplier:  Cayman Chemical 3993010 MG
Encompass_Preferred
An iridoid glycoside with diverse biological activities; reduces the proliferation of A431 cancer cells (IC50 = 310 µM); reduces the number of tumors per mouse and the percentage of mice with tumors in a mouse model of 4-nitroquinolin 1-oxide-induced carcinogenesis at 0.005% in the drinking water; decreases carrageenan-induced paw swelling in rats at 10 mg/kg; reduces blood glucose levels in a mouse model of alloxan-induced type I diabetes at 10 mg/kg
Catalog No. 50-294-3737