Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Acetylthio)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. A262125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A262125G 25 g
A26215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A262125G Supplier TCI America Supplier No. A262125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1190-93-8
Molecular Formula C4H6O3S
Molecular Weight (g/mol) 134.149
MDL Number MFCD00021763
InChI Key QSBWDKUBOZHGOU-UHFFFAOYSA-N
PubChem CID 14485
IUPAC Name 2-acetylsulfanylacetic acid
SMILES CC(=O)SCC(=O)O

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 134.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (Acetylthio)acetic Acid
RTECSNumber AF1665000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.