Learn More
StatLab 1% Acid Alcohol, 1 pint
SDP
Supplier:  StatLab AHA00188E
Encompass_Preferred
1% Acid Alcohol differentiates bacteria from background tissue by removing excess Advance Carbol Fuchsin Stain used in the MasterTech Acid Fast Bacteria Stain Kit and the Fite’s Stain Kit procedures.
Catalog No. NC1166663
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.