Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Alcohol, Strong, 5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol, Ricca Chemical
SDP

Catalog No. 26032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
260-32 1 L Natural Poly Bottle
260-16 500 mL Natural Poly Bottle
260-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 260-32 Supplier Ricca Chemical Company Supplier No. 26032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-56-1 , 7647-01-0
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236

Specifications

Physical Form Solution
CAS Min % 94.0, 4.0
CAS Max % 96.0, 6.0
Packaging Natural Poly Bottle
Color Colorless
pH <1
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol
Chemical Name or Material Acid Alcohol, Strong
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.