Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Blank, 5% (v/v) Nitric Acid, Ricca Chemical
SDP

Catalog No. PNACID51L Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PNACID51L 1 L Natural Poly Bottle
PNACID5500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PNACID51L Supplier Ricca Chemical Company Supplier No. PNACID51L
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Solubility Infinitely soluble in water
Quantity 1 L
Percent Purity 5%
Recommended Storage Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.