Learn More
Medchemexpress LLC Acid chrome blue K 100g
SDP
Supplier:  Medchemexpress LLC HYW111168100G
Encompass_Preferred
Acid chrome blue K is a metal indicator The solution color of the LAMP (Loop-mediated isothermal amplification) reaction with ACBK (Acid chrome blue K) changes from red to blue based on a decrease in the Mg2 concentration in the reaction solution[1]
Catalog No. 50-002-06422