Learn More
Parr™ Acid Digestion Bomb Replacement Cups
GSA_VASDP
Catalog No. 0102321A Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
4745 general purpose acid digestion bomb
4749 general purpose acid digestion bomb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-023-21A 4749 general purpose acid digestion bomb
01-023-2 4745 general purpose acid digestion bomb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-023-21A Supplier Parr™ Supplier No. A280AC
Add to Cart
Add to Cart
For use with Parr General Purpose Acid Digestion Bombs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Cup with Cover
Capacity (English) 0.77 oz.
Capacity (Metric) 23 mL
For Use With (Application) 4749 general purpose acid digestion bomb
For Use With (Equipment) 4749 general purpose acid digestion bomb
Material PTFE