Learn More
New Pig Acid Neutralizing Absorbent Pillow
SDP
Catalog No. 18389139 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-389-139 Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 18-389-139 Supplier New Pig Supplier No. PIL352
Add to Cart
Add to Cart
This item is not returnable. View return policy
Absorb, encapsulate and neutralize corrosives in one easy step for safer spill cleanup. Color indication identifies the best time to handle.
  • Encapsulate and neutralize the most common acid spills
  • Changes color on contact and during neutralization with liquid acids (pH below 7); color change continues until pH becomes neutral and is safe for handling
  • Instantly absorbs up to 98% concentrations of sulfuric acid
  • Gelling agents solidify acids during neutralization; prevents them from leaching out of pillow
Specifications
Material Polypropylene