Learn More
New Pig Acid-Neutralizing Spill Kit in Bucket
SDP
Catalog No. 18389124 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Acid-Neutralizing Spill Kit in Bucket
Catalog No.
18-389-124 Type: Acid-Neutralizing Spill Kit in Bucket
1 options
This item is not returnable. View return policy
Catalog No. 18-389-124 Supplier New Pig Supplier No. KIT399
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safely respond to corrosive spills with this specially equipped spill kit containing neutralizing absorbents and PPE.
  • All absorbents in this specially-designed kit neutralize acid spills
  • Changes color on contact and during neutralization with liquid acids (pH below 7); color change continues until pH becomes neutral and is safe for handling
  • Gelling agents solidify acids during neutralization; prevents them from leaching
  • Lightweight, 100% polyethylene container resists chemicals
  • Container's molded-in handles make it easy to carry Hinged polypropylene lid allows easy access and keeps contents clean, dry and secure
  • Contents are organized for quick, efficient response Includes PPE to guard against acidic materials
  • Bold lettering for quick identification
  • Temporary disposal bags with ties help make cleanup easier