Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Orange 5, TCI America™
SDP

Catalog No. A057625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A057625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A057625G Supplier TCI America Supplier No. A057625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 554-73-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.38
MDL Number MFCD00038130
InChI Key MLVYOYVMOZFHIU-UHFFFAOYSA-M
Synonym orange iv, tropaeolin oo, orangeiv, acid orange 5, diphenylamine orange, tropaeolin, unii-vr8q3r288w, tropeolin oo, orange n, solar orange iv
PubChem CID 23668839
IUPAC Name sodium 4-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Quantity 25 g
Formula Weight 375.38
Physical Form Crystalline Powder
Chemical Name or Material Acid Orange 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.