Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Orange 7 97.0+%, TCI America™
SDP

Catalog No. G0158100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0158100G 100 g
G015825G 25 g
G0158500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0158100G Supplier TCI America Supplier No. G0158100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 633-96-5
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.324
MDL Number MFCD00011657
InChI Key IGQFKZCLTLAWLO-UHFFFAOYSA-M
Synonym c.i. acid orange 7, orange ii, acid orange 7, orange, c.i. acid orange 7, monosodium salt, dsstox_cid_18881, dsstox_rid_79415, dsstox_gsid_38881, sodium 4-e-2-2-hydroxynaphthalen-1-yl diazen-1-yl benzenesulfonate, benzenesulfonic acid, 4-2-2-hydroxy-1-naphthalenyl diazenyl-, sodium salt 1:1
PubChem CID 44135675
IUPAC Name sodium;4-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonate
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 164°C
Quantity 100 g
Formula Weight 350.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acid Orange 7
EINECSNumber (5)-1455
RTECSNumber DB7084000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.