Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Acid phosphatase (Wheat Germ)

Catalog No. 89932328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-328 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-328 Supplier Abnova™ Supplier No. P5207
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 50-60kDa
Name Acid phosphatase (Wheat Germ)
Preparation Method Native protein partially purified from Wheat Germ to the 0.35-0.55 fraction by the method described by Singer, J. Biol. Chem., 174, 11 (1948)
Quantity 1 g
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name ACID
Assays Acid Phosphatase Assay
Biological Activity ≥0.15 units per mg dry weight. One Unit hydrolyzes one micromole of o-carboxyphenyl phosphate per minute at 25°C, pH 5.0.
Species Plants
Protein Tag None
Expression System Native protein partially purified from Wheat Germ to the 0.35-0.55 fraction by the method described by Singer, J. Biol. Chem., 174, 11 (1948)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.