Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Acid Sphingomyelinase Assay Kit, Fluorometric, 200 Tests

Catalog No. 502589971
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-9971 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-9971 Supplier Abcam Supplier No. AB190554200TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sensitive fluorescent assay for measuring acid sphingomyelinase activity in cell and tissue extracts.

  • Applications: Enzyme activity assay
  • Detection method: Fluorescent
  • Sample types: Cell culture extracts, tissue extracts
  • Sensitivity: 1 U/mL
  • Results type: Semi-Quantitative
  • Citation Count: 9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Acid Sphingomyelinase
For Use With (Equipment) Fluorometric Plate Reader
Format Kit
No. of Reactions 200 Tests
Substrate Properties Lipid-based Substrate
Detection Method Fluorescence
Sample Type Cell Culture Extracts, Tissue extracts
Quantity 200 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.