Learn More
Mopec Acid-Storage Cabinet, 1 Door
SDP
Catalog No. 21720002 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Door Base Cabinet
Catalog No. Product Type
21-720-002 Door Base Cabinet
1 options
Catalog No. 21-720-002 Supplier Mopec Supplier No. LC18524
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • (1) Standing Height Cabinet with leveling bolt zeroed out.
  • (1) Full-depth phenolic shelf.
  • Includes ACID sticker
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LC185-24: 24 in. L x 22 in. W x 35.1 in. H
Capacity
  • Designed for the storage of acids.
Utilities
  • Cabinets are lined with 1/8 in. thick polyethylene
  • Cabinets have a chemical-resistant 3/16 in. thick molded polyethylene lining with coved corners and a 2 in. lip at the front of the cabinet opening
Specifications
Product Type Door Base Cabinet
Color Light Gray
Material Stainless Steel