Learn More
Enzo Life Sciences Acivicin (10mg). CAS: 42228-92-2
SDP
Supplier:  Enzo Life Sciences BMLEI1130010
Encompass_Preferred
Irreversible inhibitor of γ-glutamyl transpeptidase (GGT) ID50 = 0.54 mM. Inhibits the enzymatic conversion of leukotriene C4 to leukotriene D4. Displays antitumor and anti-metastatic activity. Blocks H. Pylori GGT-induced T-cell inhibition. Inhibits bioactivation/release of nitric oxide from GSNO. Alternative name: U-42126, AT-125. Purity: ≥99% (HPLC). Solubility: Soluble in water (max. 18mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.