Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ ACK Lysing Buffer
GREENER_CHOICE

Catalog No. a1049201
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1049201 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1049201 Supplier Gibco™ Supplier No. A1049201
Add to Cart
Edge
Add to Cart

Used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow

  • Quantity: 100 mL
  • Manufactured at a cGMP-compliant facility located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Cell Culture, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lysing Buffer
Physical Form Liquid
Product Line Gibco
Quantity 100 mL
Sterility Sterile
What alternative do you offer for ACK Lysing Buffer (Cat. No. A1049201)?

Alternative products we offer are eBioscience 1X RBC Lysis Buffer (Cat. No. 00-4333-57) or eBioscience 10X RBC Lysis Buffer (Multi-species) (Cat. No. 00-4300-54).

Can I use ACK Lysing Buffer with heparinized blood?

Yes, ACK Lysing Buffer can be used with heparinized blood. The principle of lysis is osmotic treatment.

How can I use ACK Lysing Buffer to lyse red blood cells?

A volume of ACK lysing buffer corresponding to 10-20 times the volume of the blood sample is sufficient to lyse red blood cells.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.