Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACKR2 Mouse anti-Human, Clone: 1A3A2, Proteintech
SDP

Catalog No. 501736205 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6205 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6205 Supplier Proteintech Group Inc Supplier No. 600461IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.

Specifications

Antigen ACKR2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A3A2
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACKR2
Gene Accession No. O00590
Gene Alias ACKR2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6
Gene Symbols ACKR2
Host Species Mouse
Immunogen ACKR2 Fusion Protein Ag1941
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.