Learn More
Sino Biological AcMNPV AcMNPV-GP64 PAb 100UL 40496-T62-100
Supplier:  Sino Biological 40496T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 ( Catalog#40496-V08B; NP_054158.1; Met1-Thr481). AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 specific IgG was purified by AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 affinity chromatography.
Catalog No. 50-240-9187
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.