Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AcmNPV gp64 Monoclonal Antibody (M001)
GREENER_CHOICE

Catalog No. PIMA529952 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529952 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529952 Supplier Invitrogen™ Supplier No. MA529952
  • $523.00
    $436.65 / Each of 1

    Save $86.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for AcmNPV Envelope glycoprotein gp64/AcmNPV-gp64.

Class III viral fusion protein. Envelope phosphoglycoprotein which mediates the fusion of viral and host endosomal membranes leading to virus entry into the host cell. After receptor-mediated internalization of the virus, gp64 undergoes conformational change into a fusion-competent state at low pH, and the nucleocapsid is released into the cytoplasm after cell fusion. May also play role in budding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AcmNPV gp64
Applications Neutralization
Classification Monoclonal
Clone M001
Concentration 10.02 mg/mL
Conjugate Unconjugated
Formulation Arginine, histidine with 0.02% Tween 80, 120mM NaCl and no preservative; pH 6.0
Gene Alias Gp64
Host Species Mouse
Immunogen Recombinant Autographa californica nucleopolyhedrovirus (AcNPV)(strain E2).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.