Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aconitate hydratase B Mouse anti-E. coli, HRP, Antibody Pair, Novus Biologicals™

Antibody pair for Solid phase sandwich ELISA
$458.00 - $1124.00
Specifications
Antigen | Aconitate hydratase B |
---|---|
Applications | ELISA |
Classification | Monoclonal |
Conjugate | HRP |
Host Species | Mouse |
Description
Solid phase sandwich ELISA for quantitative determination of E. coli Aconitate hydratase B.Specifications
Aconitate hydratase B | |
Monoclonal | |
Mouse | |
E. coli | |
Storage of components varies. See protocol for specific instructions. |
ELISA | |
HRP | |
RUO | |
acnB, hypothetical protein, yacI, yacJ | |
Antibody Pairs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title