Learn More
Cayman Chemical AcoramIdIshydrochlorIde 25mg
SDP
Supplier:  Cayman Chemical 4153625 MG
Encompass_Preferred
A multi-tail ionizable cationic phospholipid has been used in combination with other lipids in the formation of LNPs LNPs containing PL1 and encapsulating mRNA encoding CD137 or OX40 increase T cell antitumor immunity in various mouse xenograft models
Catalog No. 50-294-5587
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.