Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACPP Mouse anti-Human, Rat, Clone: 1G2C9, Proteintech
SDP

Catalog No. p-7212523 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6280 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6280 Supplier Proteintech Group Inc Supplier No. 601761IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.

Specifications

Antigen ACPP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G2C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACPP
Gene Accession No. P15309, P20646
Gene Alias acid phosphatase, prostate, ACP 3, ACP3, ACPP, PAP, Prostatic acid phosphatase
Gene Symbols ACPP
Host Species Mouse
Immunogen ACPP Fusion Protein Ag8578
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55, 56780
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.