Learn More
Waters Corporation ACQUITY UPLC BEH450 SEC Guard Column
Shop All Waters Chromatography Solutions Products
Description
- Based upon the highly successful BEH particle technology, the ACQUITY UPLC BEH450 SEC 2.5 μm column is fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 100,000 to 1,500,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.
Specifications
Specifications
| Quantity | 1/Ea. |
| Particle Size | 2.5 μm |
| Length (Metric) | 30 mm |
| Pore Size | 450 Å |
| Type | Guard Column |
| Diameter (Metric) | 4.6 mm |
| For Use With (Application) | Protein |
| Particle Shape | Spherical |
| Endcapped | No |
| USP Type | L33 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.