Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine-9-Carboxylic Acid, MP Biomedicals™

Catalog No. MP520111401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520111401 1 g
MP52011142 250 mg
MP520111405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP520111401 Supplier MP Biomedicals, Inc Supplier No. 0520111401

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Acridine-9-Carboxylic Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332927-03-4
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.23
MDL Number MFCD00149578
InChI Key IYRYQBAAHMBIFT-UHFFFAOYSA-N
IUPAC Name acridine-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 300.6°C to 304.9°C
Color Green-yellow
Quantity 1 g
Physical Form Powder
Assay >97%
storageNote1 +4°C, protect from light, store under nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.