Learn More
Apexbio Technology LLC Acridine Orange Staining Kit 100T
SDP
Supplier:  Apexbio Technology LLC K2251100
Encompass_Preferred
Acridine Orange Staining Kit is a fluorescence-based nucleic acid detection reagent designed for distinguishing cellular DNA and RNA particularly in apoptosis assays Acridine Orange is a cell membrane-permeant dye that selectively binds to double-stranded DNA emitting green fluorescence and to RNA or single-stranded DNA emitting orange or red fluorescence It is frequently used in combination with cell-impermeable dyes such as Ethidium Bromide or Propidium Iodide to differentiate viable apoptotic and necrotic cells according to selective membrane permeability and nuclear morphology changes during apoptosis Based on these properties Acridine Orange Staining Kit holds research potential in apoptosis detection and cell viability analysis
Catalog No. 50-000-97045